Order Entry
Austria
Orders LinkContactUsLinkComponent
Spoon spatula

Spoon spatula

190 mm, 12 mm, 20×25 mm
Catalog # 231-0111
Supplier:   NICKEL ELECTRO
Spoon spatula
Catalog # 231-0111
Supplier:  NICKEL ELECTRO
Spoon spatula
Spoon spatula

190 mm, 12 mm, 20×25 mm

Price
16,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Double spatula with offset spoon end and scraper end.

  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning
Specifications
  • Length:
    190 mm
  • Blade width:
    12 mm
  • Spoon size:
    20×25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.