Orders LinkContactUsLinkComponent
Spoon spatulas
 :   BOCHEM
Spoon spatulas
 :  BOCHEM
Spoon spatulas
Spoon spatulas
Stainless steel. Universal spoon spatula designed for a wide range of uses.
  • One end is shaped like a spoon and the other end like a spatula
  • With rounded handle
  • High thermal resistance, autoclavable
  • Excellent resistance to chemicals
  • Quick and easy to clean
Product Family Options
  • Length
    Spoon size
  • 130 mm
    10×5 mm
    1 * 1 ST
    1 * 1 ST - 1,90 €
    1,90 €
    1 * 1 ST
       1,90 €
  • 185 mm
    10×5 mm
    1 * 1 ST
    1 * 1 ST - 2,10 €
    2,10 €
    1 * 1 ST
       2,10 €
  • 210 mm
    12×5 mm
    1 * 1 ST
    1 * 1 ST - 2,30 €
    2,30 €
    1 * 1 ST
       2,30 €
  • 180 mm
    25×8 mm
    1 * 1 ST
    1 * 1 ST - 2,40 €
    2,40 €
    1 * 1 ST
       2,40 €
  • 210 mm
    25×12 mm
    1 * 1 ST
    1 * 1 ST - 2,40 €
    2,40 €
    1 * 1 ST
       2,40 €
  • 230 mm
    25×12 mm
    1 * 1 ST
    1 * 1 ST - 2,60 €
    2,60 €
    1 * 1 ST
       2,60 €