Order Entry
Austria
ContactUsLinkComponent
Signs, prohibition sign, indoor and outdoor, kroschke
Signs, prohibition sign, indoor and outdoor, kroschke
Catalog # 116-2033
Supplier:  KROSCHKE
undefined
Signs, prohibition sign, indoor and outdoor, kroschke
Catalog # 116-2033
Supplier:  KROSCHKE

Specifications

  • Description:
    Food and drink prohibited
  • Material:
    PVC
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

These prohibition signs are made from self-adhesive PVC foil and proscribe practices that could be dangerous. According to VBG 125 these must be affixed visibly and permanently at all times.

  • Suitable for indoor and outdoor use
  • Visible from a distance of approx. 8 m

Ø 200 mm

Certifications: VBG 125 (BGV A8)