ESD occupational shoes, lace-up, X-Light, 7131133/7131138
Lieferant: Abeba
Zertifikate
Über diesen Artikel
ESD shoe made from smooth leather, reflective material, and a lightweight PU walking sole, resistant to hydrocarbons.
- Perspiration-absorbent special lining with silver ions
- Replaceable active comfort insole provides optimised comfort
- orthopaedic insoles possible
- Sanitized® treatment reduces odour and bacterial growth
- Lightweight PU sole with high slip resistance and shock-absorbing system
Colour: White or black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O2, FO, SRC.
Zugehörige Informationen
Frequently Bought Together