ContactUsLinkComponent
Sealing tape for MicroWell™ plates, Nunc™
Sealing tape for MicroWell™ plates, Nunc™
  732-2610
   Low Manufacturing Impact
undefined
Sealing tape for MicroWell™ plates, Nunc™
  732-2610
   Low Manufacturing Impact
  • Description:
    Sealing tape, polyester
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    200
Polyester film with acrylic adhesive. Provides an effective seal for all MicroWell™ plates used in PCR and cell culture.
  • Resistant to temperatures between −70 and +100 °C
  • Minimum evaporation and well-to-well cross-contamination
Dimensions: 134×80 mm
Dimensions: 134×80 mm