Orders LinkContactUsLinkComponent
Scoops
Scoops
Supplier:   VITLAB
Scoops
Supplier:  VITLAB
Certificates
Scoops
8 Options Available Below
Product Details & Documents
HDPE, translucent. With flat ends and reinforced handle.
  • Good chemical resistance
  • Heat resistant up to +105 °C
  • The capacity of the scoop is stamped into the handle for easy reading
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 15 ml
    115 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 9,90 €
    9,90 €
    1 * 12 ST
       9,90 €
  • 25 ml
    135 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 13,80 €
    13,80 €
    1 * 12 ST
       13,80 €
  • 65 ml
    185 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 19,40 €
    19,40 €
    1 * 12 ST
       19,40 €
  • 110 ml
    215 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 23,60 €
    23,60 €
    1 * 12 ST
       23,60 €
  • 150 ml
    250 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 26,90 €
    26,90 €
    1 * 12 ST
       26,90 €
  • 350 ml
    310 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 23,30 €
    23,30 €
    1 * 6 ST
       23,30 €
  • 750 ml
    350 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 38,50 €
    38,50 €
    1 * 6 ST
       38,50 €
  • 1250 ml
    400 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 53,00 €
    53,00 €
    1 * 6 ST
       53,00 €