Orders LinkContactUsLinkComponent
Ladle scoops
Supplier:   BOCHEM
Ladle scoops
Supplier:  BOCHEM
Certificates
Ladle scoops
6 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic and electropolished.
  • Complete with flat handle
  • Corrosion resistant
  • Excellent thermal resistance
Product Family Options
Product Information
  • Capacity
    Length
    Ø
    Availability
    Price
  • 90 ml
    185 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,90 €
    15,90 €
    1 * 1 ST
       15,90 €
  • 200 ml
    315 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,20 €
    19,20 €
    1 * 1 ST
       19,20 €
  • 220 ml
    315 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,00 €
    26,00 €
    1 * 1 ST
       26,00 €
  • 620 ml
    345 mm
    140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,10 €
    41,10 €
    1 * 1 ST
       41,10 €
  • 1000 ml
    350 mm
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,70 €
    77,70 €
    1 * 1 ST
       77,70 €
  • 1400 ml
    350 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €