Orders LinkContactUsLinkComponent
Thread and vessel scissors
Thread and vessel scissors
Thread and vessel scissors
Thread and vessel scissors
Stainless steel 18/10, anti-magnetic.
  • Sharp-blunt
  • Ideal for a wide range of laboratory applications
  • Acid-resistant
  • Autoclavable
Product Family Options
  • Type
    Length
  • Straight, sharp/blunt
    110 mm
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • Straight, sharp/blunt
    130 mm
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • Straight, sharp/blunt
    145 mm
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • Straight, sharp/blunt
    160 mm
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €