VWR®, Laboratory Jacks
Supplier: VWR Collection
Certificates
About this item
High quality, stable and inexpensive range of lifting platforms for a variety of uses. With plates made from matt, brushed, non rusting stainless steel, and smooth running aluminium frame with scissor action. Rubber feet ensure stable support.
- Ergonomically-designed height adjustment knob for easy operation
- Available with several different platform sizes
- M10 hole for retort rod (except 245-1220)
Frequently Bought Together