Flat flange lids with 5 necks
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With flat laboratory flange (DN). Straight middle neck, three angled side necks and one straight side neck with standard ground joints.
Frequently Bought Together