Reaction vessels, cylindrical, with flat flange, thermal jacket and drain valve
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With easily readable graduation and flat laboratory flange (LF) with groove. With two connectors with GL 14/GL 18 or KF 15/KF 25 ball flanges.
Frequently Bought Together