Order Entry
Austria
ContactUsLinkComponent
Beta radiation bin, Nalgene®
Beta radiation bin, Nalgene®
Catalog # 113-2806
   Low Manufacturing Impact
undefined
Beta radiation bin, Nalgene®
Catalog # 113-2806
   Low Manufacturing Impact

Specifications

  • Description:
    Waste Bin for 2 litre
  • Dimensions:
    140×286 mm (Ø×H)
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1

Specifications

About this item

Acrylic, transparent, thickness: 9 mm.

  • HDPE 2 litre bottle for liquid waste is included; organic solvents may require a different bottle
  • Shield encases the waste bottle without obstructing access
Delivery: Supplied with PE bottle for liquid waste.