Test tube racks
Supplier: KARTELL
Certificates
About this item
PP, coloured.
- Excellent chemical resistance
- Heat resistant up to +121 °C, autoclavable
- Very stable, robust
Versatile and good value. The racks do not float, so they are ideal for use in water baths. Alphanumeric position designators for easy sample identification. The racks are delivered dismantled and can be put together securely and permanently in a few steps, without tools.
Frequently Bought Together