Microtube racks, floating, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PP, floating.
- Temperature resistant from −70 to +100 °C, autoclavable
- Alphanumeric labelling of rack positions for easy identification of samples
- Handles enable the racks to be carried, shifted and removed easily
Ideal for thawing, cooling or incubating samples at low or high temperatures. Racks float even when full. Tubes are kept below the rim. Round racks 211-7313 and 213-7314 fit in 400 and 1000 ml low-form beakers.
Specifications
- Catalog No:
- 211-7313
- 211-7312
- 211-7314
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)