ContactUsLinkComponent
Cleaner for spectrophotometer cells, Hellmanex® III
Cleaner for spectrophotometer cells, Hellmanex® III
Catalog # 634-0666
Supplier:  HELLMA
undefined
Cleaner for spectrophotometer cells, Hellmanex® III
Catalog # 634-0666
Supplier:  HELLMA
Specifications
  • Description:
    Hellmanex III
  • Environmentally Preferable:
  • Pk:
    1 L
Specifications
About this item
An alkaline liquid concentrate, which is typically diluted to 2% with water for cleaning quartz and glass cells. It can also be used to clean other sensitive optical components made of glass, quartz, sapphire and porcelain.
Hellmanex® III is a complete replacement for chromo sulphuric acid. It removes even the most stubborn contaminants such as fats, waxes, and proteins whilst being considerably less hazardous and corrosive than some of the traditional alternatives.