Order Entry
Austria
ContactUsLinkComponent
Rack for cryotubes
Rack for cryotubes
Catalog # GREI802501_5
Supplier:  Greiner Bio-One
undefined
Rack for cryotubes
Catalog # GREI802501_5
Supplier:  Greiner Bio-One

Specifications

  • For:
    Ø 12,5 mm
  • No. of Holes:
    40 (4×10)
  • W×D×H:
    200×100×22 mm
  • Pk:
    5

Specifications

About this item

PC, blue.

  • Non slip rubber base

This rack is suitable for cryotubes with a 'starfoot' base and allows one-handed manipulation of the tubes.