Orders LinkContactUsLinkComponent
Horizontal developing chambers
Supplier:   CAMAG
Horizontal developing chambers
Supplier:  CAMAG
Certificates
Horizontal developing chambers
Horizontal developing chambers
2 Options Available Below
Product Details & Documents
Horizontal developing chambers allows the number of samples to be doubled when compared to a vertical tank. Plates are developed from opposite sides simultaneously towards the middle.
  • Choice of two configurations: sandwich or tank
  • Suitable for all types of solvents
  • Available in two plates sizes
Product Family Options
Product Information
  • Availability
    Price
  • 552-0047
    Chamber for 100×100 mm plates
    Chamber for 100×100 mm plates
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 210,00 €
    1 210,00 €
    1 * 1 ST
       1 210,00 €
  • 552-0048
    Chamber for 200×100 mm plates
    Chamber for 200×100 mm plates
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €