Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pipette Tips
Tip refill systems, Axygen®
Tip refill systems, Axygen®
:
Corning
Tip refill systems, Axygen®
:
Corning
Transparent PP, autoclavable.
Easy to load refill racks
Certified RNase-, DNase- and endotoxin-free
Universal fit tips have a bevelled reference and graduated marks
CoC_AXYGRFL-300-C_06522326
Empty refill boxes with lid