Orders LinkContactUsLinkComponent
Pasteur pipettes, PE
Pasteur pipettes, PE
Pasteur pipettes, PE
Supplier:  Alpha Laboratories
Certificates
Pasteur pipettes, PE
Pasteur pipettes, PE
3 Options Available Below
Product Details & Documents
These single-use transfer pipettes are unbreakable, don't require a separate bulb and, as they are manufactured from soft PE plastic, can be recycled. Sterile.
612-3751 for accessing deep vessels such as Winchesters and tissue culture flasks.
612-1793: type C, with graduations at 0,25 ml.
612-3751: type E, without graduations.
612-3752: type D, with graduations at 0,5 ml.
Packaging: 20 pipettes/bag.
Product Family Options
Product Information
  • Capacity
    Drop vol.
    Bulb draw
    Length
    Graduation
    Sterile
    Packed
    Availability
    Price
  • 1,0 ml
    40 µl
    3,4 ml
    155 mm
    0,25 ml
    +
    20/bag
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 74,60 €
    74,60 €
    1 * 500 ST
       74,60 €
  • -
    40 µl
    2,3 ml
    225 mm
    - ml
    +
    20/bag
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 123,00 €
    123,00 €
    1 * 400 ST
       123,00 €
  • 3,0 ml
    50 µl
    3,2 ml
    155 mm
    0,5 ml
    +
    20/bag
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 85,20 €
    85,20 €
    1 * 500 ST
       85,20 €