Orders LinkContactUsLinkComponent
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
Supplier:   GLW
Sterilisation Boxes for Pipettes
Supplier:  GLW
Certificates
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
8 Options Available Below
Product Details & Documents
GLW Pipette Boxes are made from aluminum. Due to its square-shaped form, the box remains stationary while working and will not roll away as is the case with round containers. The box is available in four different sizes.
  • Can be autoclaved at +121 °C; can withstand temperatures up to +250 °C
  • 'Economy' boxes: without silicone
  • 'Premium' boxes: integral silicone coating in base and lid
  • W×D (mm): 68×68
W×D (mm): 68×68
Product Family Options
'Economy' boxes, without silicone
Product Information
  • W×D×H
    Availability
    Price
  • 612-2085
    Square base
    Square base
    68×68×150/240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,50 €
    30,50 €
    1 * 1 ST
       30,50 €
  • 612-2086
    Square base
    Square base
    68×68×310/400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,54 €
    31,54 €
    1 * 1 ST
       31,54 €
  • 612-2089
    Square base
    Square base
    68×68×230/320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,76 €
    30,76 €
    1 * 1 ST
       30,76 €
  • 612-2090
    Square base
    Square base
    68×68×310/450 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,14 €
    39,14 €
    1 * 1 ST
       39,14 €
'Premium' boxes, with silicone
Product Information
  • W×D×H
    Availability
    Price
  • 612-2087
    Square base
    Square base
    68×68×150/240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,14 €
    36,14 €
    1 * 1 ST
       36,14 €
  • 612-2088
    Square base
    Square base
    68×68×310/400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,70 €
    37,70 €
    1 * 1 ST
       37,70 €
  • 612-2091
    Square base
    Square base
    68×68×230/320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,92 €
    36,92 €
    1 * 1 ST
       36,92 €
  • 612-2092
    Square base
    Square base
    68×68×310/450 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,42 €
    45,42 €
    1 * 1 ST
       45,42 €