Order Entry
Austria
ContactUsLinkComponent
Storage and organisation system
Storage and organisation system
  612-1861
 :  HEATHROW
undefined
Storage and organisation system
  612-1861
 :  HEATHROW

 

  • Colour:
    Blue
  • Description:
    Manual pipette rack
  • W×D×H:
    88×291×401 mm
  • Pk:
    1

 

 

ABS, coloured. For standard and tissue culture pipettes, and pipette boxes. Strong, stable construction.

  • Suitable for standard and tissue culture pipettes from 0,1 to 25 ml
  • Four compartments with side viewing windows allow a clear view, even when several systems are arranged next to each other
  • Lateral clamp holders for pipetting aids or pipetting bulbs