Order Entry
Austria
ContactUsLinkComponent
 
Pipette stand, rotary
Catalog # 612-4008
Supplier:  KARTELL
undefined
Pipette stand, rotary
Catalog # 612-4008
Supplier:  KARTELL

Specifications

  • Ø×H:
    230×470 mm
  • Colour:
    Red
  • Description:
    Pipette holder, rotary
  • Pk:
    1

Specifications

About this item

Rotary pipette holder made from PP, for up to 94 pipettes.

  • Rotating base
  • Accommodates up to 94 pipettes with a diameter not exceeding 14 mm

Ø×H: 230×470 mm