Orders LinkContactUsLinkComponent
Pipette controller, pipetus®-junior
Pipette controller, pipetus®-junior
pipetus®-junior complete
Catalog # 612-2604
Supplier:   HIRSCHMANN
Pipette controller, pipetus®-junior
Catalog # 612-2604
Supplier:  HIRSCHMANN
Pipette controller, pipetus®-junior
Pipette controller, pipetus®-junior
pipetus®-junior complete
Price
160,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Pipette controller for glass and plastic pipettes from 0,5 to 25 ml.
  • Easy, ergonomic, safe and accurate
  • Total expulsion of the liquid simply by pressing the button on the back of the instrument
  • The integrated, hydrophobic PTFE membrane filter ensures the pipette is protected against accidental overfilling
Full pressure on the fill button allows suction of around 2 ml. By pressing on the empty button, the pipette is emptied gravimetrically and the degree of pressure on the button determines the rate of flow.
Specifications
  • Description:
    pipetus®-junior complete
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Pipette holder, complete, for pipetus® junior (units manufactured until May 2004, up to serial number 4404001)
    Pipette holder, complete, for pipetus® junior (units manufactured until May 2004, up to serial number 4404001)
    612-2612
    Log in for availability
    1 * 1 ST - 50,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Spacer
    Spacer
    612-2617
    Log in for availability
    1 * 1 ST - 8,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (5 filters)
    Round filter set with one-sided connection (5 filters)
    612-1982
    Log in for availability
    1 * 5 ST - 40,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (100 filters)
    Round filter set with one-sided connection (100 filters)
    HIRS9903706
    Log in for availability
    1 * 100 ST - 601,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone adapter
    Silicone adapter
    612-2615
    Log in for availability
    1 * 1 ST - 9,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (50 filters)
    Round filter set with one-sided connection (50 filters)
    HIRS9903705
    Log in for availability
    1 * 50 ST - 347,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette holder, complete, for pipetus® junior (units manufactured from May 2004, beginning with serial number 4405001)
    Pipette holder, complete, for pipetus® junior (units manufactured from May 2004, beginning with serial number 4405001)
    HIRS9902400
    Log in for availability
    1 * 1 ST - 50,10 €