Orders LinkContactUsLinkComponent
Pipette controller, pipetus®
Pipette controller, pipetus®
pipetus®, EU/UK/US-plugs
Catalog # 612-1874
Supplier:   HIRSCHMANN
Pipette controller, pipetus®
Catalog # 612-1874
Supplier:  HIRSCHMANN
Pipette controller, pipetus®
Pipette controller, pipetus®
pipetus®, EU/UK/US-plugs
Price
490,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Pipette controller for glass and plastic pipettes from 0,1 to 200 ml.
  • Unique and ecologically sound with up to 50 hours of independent running, cadmium-free NiMH batteries
  • Coloured display allows visual check of the battery charging status, the selected pipetting mode (Ex or blow-out), and the selected speed
  • Safe thanks to the safety return valve and highly sensitive valves for precise meniscus adjustment
  • Silent due to vibrationless pump motor
  • Silicone adapter for entirely leakproof positioning of each pipette
This pipette has a hydrophobic PTFE membrane filter with a porosity of 0,2 µm, intelligent charging technology, no memory effect, and protection against overcharging the batteries ensured by microprocessor control. The 'Speed' key allows you to choose the ideal pipetting speed from the five speeds offered, with colour indicator.
Ordering information: Supplied complete with charger and plate, two PTFE filters, one safety valve and one wall bracket.
Documents
Specifications
  • Description:
    pipetus®, EU/UK/US-plugs
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (5 filters)
    Round filter set with one-sided connection (5 filters)
    612-1982
    Log in for availability
    1 * 5 ST - 40,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (100 filters)
    Round filter set with one-sided connection (100 filters)
    HIRS9903706
    Log in for availability
    1 * 100 ST - 601,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mains-operated charger unit with charging stand, EU-plug
    Mains-operated charger unit with charging stand, EU-plug
    612-2600
    Log in for availability
    1 * 1 ST - 97,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (50 filters)
    Round filter set with one-sided connection (50 filters)
    HIRS9903705
    Log in for availability
    1 * 50 ST - 347,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette holder, complete, white, for pipetus®/pipetus® accu/pipetus® classic
    Pipette holder, complete, white, for pipetus®/pipetus® accu/pipetus® classic
    612-1875
    Log in for availability
    1 * 1 ST - 69,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Wall holder, for all pipetus® units
    Wall holder, for all pipetus® units
    612-4199
    Log in for availability
    1 * 1 ST - 21,10 €