Orders LinkContactUsLinkComponent
Pipette controller, Pi-Pump
Pipette controller, Pi-Pump
Supplier:   KARTELL
Pipette controller, Pi-Pump
Supplier:  KARTELL
Certificates
Pipette controller, Pi-Pump
Pipette controller, Pi-Pump
2 Options Available Below
Product Details & Documents
Pipetting aid for all plastic and glass pipettes.
  • Suction and dispensing achieved by rotating the wheel
  • Liquid evacuated by pressing the lever
Product Family Options
Product Information
  • Availability
    Price
  • 612-1926
    Pipette controller Pi-Pump, green, for 10 ml pipettes
    Pipette controller Pi-Pump, green, for 10 ml pipettes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,90 €
    37,90 €
    1 * 1 ST
       37,90 €
  • 612-1927
    Pipette controller Pi-Pump, red, for 25 ml pipettes
    Pipette controller Pi-Pump, red, for 25 ml pipettes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,60 €
    24,60 €
    1 * 1 ST
       24,60 €