Orders LinkContactUsLinkComponent
Micro pipettes, minicaps®
Micro pipettes, minicaps®
Micro pipettes, minicaps®
Supplier:  HIRSCHMANN
Certificates
Micro pipettes, minicaps®
Micro pipettes, minicaps®
14 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, not heparinised, disposable.
  • Conformity certified
  • End to end: contains the stated volume when filled from one end to the other
  • Safe packaging for protection during transportation
DIN ISO 7550, IVD 98/79 EC
Accuracy: 0,5%
Reproducibility: 1%
Certifications: DIN ISO 7550, IVD 98/79 EC
Packaging: 100 micro pipettes are packed in a glass vial with colour-coded dispenser cap, 10 of these vials are packed into a stable carton box.
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 25 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 100 µl
    100 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 1 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 2 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 5 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 10 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 40 µl
    30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 0,5 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 3 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 4 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 20 µl
    30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 30 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 50 µl
    32 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 6,66 µl
    72 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €