Graduated pipettes, type 3, Class AS, total delivery
AR-Glas®.
- Calibrated to deliver 'Ex', certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
- Marks and inscriptions in amber stain, graduated to the tip
- From 5 ml with a cotton plug as standard
- Waiting time five seconds
DIN EN ISO 835
Length: 360 mm ±5 mm except for 25 and 50 ml pipettes: 450 mm ±5 mm
: DIN EN ISO 835. Conformity certified (DE-M marking).
Frequently Bought Together