Orders LinkContactUsLinkComponent
Mortars, without pestles
Mortars, without pestles
Mortars, without pestles
Certificates
Mortars, without pestles
Mortars, without pestles
22 Options Available Below
Product Details & Documents
Porcelain with lip. Choice of glazed or unglazed grinding surface.
Certifications: Manufactured to DIN 12906.
Ordering information: Pestle must be ordered separately.
Product Family Options
Glazed
Product Information
  • Capacity
    Height
    Ø int.
    Availability
    Price
  • 20 ml
    32 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,80 €
    34,80 €
    1 * 1 ST
       34,80 €
  • 70 ml
    40 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,80 €
    42,80 €
    1 * 1 ST
       42,80 €
  • 75 ml
    45 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,50 €
    44,50 €
    1 * 1 ST
       44,50 €
  • 100 ml
    50 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,70 €
    52,70 €
    1 * 1 ST
       52,70 €
  • 170 ml
    55 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,90 €
    54,90 €
    1 * 1 ST
       54,90 €
  • 250 ml
    63 mm
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,30 €
    66,30 €
    1 * 1 ST
       66,30 €
  • 400 ml
    65 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 79,80 €
    79,80 €
    1 * 1 ST
       79,80 €
  • 650 ml
    70 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • 1000 ml
    80 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 250,00 €
    250,00 €
    1 * 1 ST
       250,00 €
  • 2400 ml
    100 mm
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 554,00 €
    554,00 €
    1 * 1 ST
       554,00 €
  • 6000 ml
    140 mm
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 709,00 €
    709,00 €
    1 * 1 ST
       709,00 €
Unglazed
Product Information
  • Capacity
    Height
    Ø int.
    Availability
    Price
  • 20 ml
    32 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,90 €
    15,90 €
    1 * 1 ST
       15,90 €
  • 70 ml
    40 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,00 €
    24,00 €
    1 * 1 ST
       24,00 €
  • 75 ml
    45 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,60 €
    25,60 €
    1 * 1 ST
       25,60 €
  • 100 ml
    50 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,90 €
    33,90 €
    1 * 1 ST
       33,90 €
  • 170 ml
    55 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • 250 ml
    63 mm
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,40 €
    47,40 €
    1 * 1 ST
       47,40 €
  • 400 ml
    65 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,90 €
    60,90 €
    1 * 1 ST
       60,90 €
  • 650 ml
    70 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,30 €
    92,30 €
    1 * 1 ST
       92,30 €
  • 1000 ml
    80 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €
  • 2400 ml
    100 mm
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 417,00 €
    417,00 €
    1 * 1 ST
       417,00 €
  • 6000 ml
    140 mm
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 698,00 €
    698,00 €
    1 * 1 ST
       698,00 €