Orders LinkContactUsLinkComponent
Mop systems, Edgeless®
Mop systems, Edgeless®
Supplier:   CONTEC
Mop systems, Edgeless®
Supplier:  CONTEC
Certificates
Mop systems, Edgeless®
Mop systems, Edgeless®
3 Options Available Below
Product Details & Documents
Designed for wet mopping in controlled environments and cleanrooms for effective cleaning and retrieval of excess fluid. Ideal for use in sterile environments. Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption.
  • Mop heads are free from any cellulose contaminants
  • Stainless bucket, downpress wringer and insert maximise wringing pressure and effectiveness
  • All system components and buckets are compatible with gamma, ETO, and autoclave sterilisation
Mop heads are laundered and packaged in a ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Ordering information: All mop system components must be ordered separately. Plastic bucket systems include cart, buckets, wringer, and casters. Stainless steel bucket systems include trolley, buckets, wringer, and casters.
Product Family Options
Product Information
  • Dimensions
    Length
    Capacity
    Availability
    Price
  • CONTEL-MOPIR
    Standard edgeless mop head, sterile
    Standard edgeless mop head, sterile
    -
    -
    -
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 1 410,00 €
    1 410,00 €
    1 * 20 ST
       1 410,00 €
  • 115-0174
    Standard edgeless mop head, non sterile
    Standard edgeless mop head, non sterile
    -
    -
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,00 €
    62,00 €
    1 * 1 ST
       62,00 €
  • CONT2646F
    Edgeless floor mop frame QuickConnect™, polypropylene copolymer frame with stainless steel connector ; compatible with all styles of Edgeless mops and easily attaches to QuickConnect handles. Autoclavable at 121°C for 30 min.
    Edgeless floor mop frame QuickConnect™, polypropylene copolymer frame with stainless steel connector ; compatible with all styles of Edgeless mops and easily attaches to QuickConnect handles. Autoclavable at 121°C for 30 min.
    150×180 mm
    -
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 193,00 €
    193,00 €
    1 * 1 ST
       193,00 €