Orders LinkContactUsLinkComponent
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
New Product
Supplier:   Texwipe
TechniCloth® Flat Mop Covers
Supplier:  Texwipe
New Product
Certificates
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
2 Options Available Below
Product Details & Documents
Mop covers (ISO Class 3 - 8) for cleaning and decontaminating floors.
  • Made without chemical binders or additives
  • Non-woven, economical polyester/cellulose for cleaning and removing residues
  • Chemically resistant, withstand exposure to common cleaning and disinfecting solutions
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • TEXWTX630
    Technicloth® floor mop wipers
    Technicloth® floor mop wipers
    250×600 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 1 037,86 €
    1 037,86 €
    1 * 12 ST
       1 037,86 €
    Specifications:
    New Product
    New Product
  • 115-0480
    Technicloth® floor mop wipers
    Technicloth® floor mop wipers
    690×380 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 462,06 €
    462,06 €
    1 * 4 ST
       462,06 €
    Specifications:
    New Product
    New Product