ContactUsLinkComponent
Mops, PilloMop™
Mops, PilloMop™
  MICRPM-1_I
 :  MICRONOVA
undefined
Mops, PilloMop™
  MICRPM-1_I
 :  MICRONOVA
  • Description:
    Mop head, PVA, irradiated
  • Seq:
    01
  • Pk:
    1
These flat mop heads move easily, ideal for reaching awkward corners, shelving, and narrow chambers. (W×D) 229×127 mm. The slip cover system offers precise manual wall wiping control. Polyester base pads measure (W×D) 203×102 mm. PilloMop™ adapter is required to attach all mop heads to stainless steel universal handle.
  • The polyester slip cover fits over the base pad and can be changed with each wipe
  • System can be laundered, is autoclavable, and reusable
  • PVA covering designed for semiconductor, electronics applications requiring low-linting absorbence
  • Autoclavable two-ply polyester foam cover is low-linting for critical and sterile applications
  • PilloMop™ lite an absorbent latex/rayon blend, can be used with strong disinfectants and be autoclaved
 : All components must be ordered separately.