Orders LinkContactUsLinkComponent
Mops, MicroMop™
   Sustainable Options Available
Supplier:   MICRONOVA
Mops, MicroMop™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Mops, MicroMop™
3 Options Available Below
Product Details & Documents
MicroMop™ are ideal for quick and efficient cleaning of large surface areas. Suitable for wafer processing, electronics, pharmaceutical, and biotech applications. All mop heads attach to stainless steel universal handle via the orange polypropylene quick disconnect adapter. For less critical applications, choose the fibreglass/polypropylene handle with attached adapter.
  • Low-linting, quick drying PVA is ideal for high-tech applications. Can be laundered
  • Polyester has good wicking capability, can be autoclaved and used with strong disinfectants
  • Lite™ is sturdy latex/rayon blend for controlled environments and cleanroom support areas; mop head can be autoclaved for medical and sterile applications
 : All components must be ordered separately.
Specifications for Product Family
Specifications
  • Catalog No:
  • 129-0506
  • 129-0234
  • MICRMHC-1
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
Specifications
Product Family Options
Mop heads
Product Information
  • Length
    Availability
    Price
  • 129-0506
    Mop head, NovaLite™
    Mop head, NovaLite™
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 197,00 €
    197,00 €
    1 * 1 ST
       197,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0506
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 129-0234
    Mop head, polyester
    Mop head, polyester
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,30 €
    84,30 €
    1 * 1 ST
       84,30 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0234
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Handles
Product Information
  • Length
    Availability
    Price
  • MICRMHC-1
    MicroMop® handle, Fibreglass
    MicroMop® handle, Fibreglass
    1524 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,20 €
    72,20 €
    1 * 1 ST
       72,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    MICRMHC-1
    Environmentally Preferable
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    Log in for availability
    1 * 1 ST - 157,00 €