Orders LinkContactUsLinkComponent
Slide mailer
Slide mailer
Slide mailers, Translucent, PP, 43×24×88 mm, 5
Catalog # 631-9782
Supplier:   HEATHROW
Slide mailer
Catalog # 631-9782
Supplier:  HEATHROW
Slide mailer
Slide mailer
Slide mailers, Translucent, PP, 43×24×88 mm, 5
Price
23,40 €
Log in for availability
1 * 25 ST
Product Details & Documents
This slide mailer is specially useful for the transportation and storage of microscopy slides.
  • Snap closure on one end
  • Guide rails on each slide
  • For five standard slides
Documents
Specifications
  • No. of slides:
    5
  • Colour:
    Translucent
  • Description:
    Slide mailers
  • Material:
    PP
  • W×D×H:
    43×24×88 mm