Orders LinkContactUsLinkComponent
Slide holders, SlideFolder™
Slide holders, SlideFolder™
Supplier:   Simport Scientific
Slide holders, SlideFolder™
Supplier:  Simport Scientific
Certificates
Slide holders, SlideFolder™
Slide holders, SlideFolder™
7 Options Available Below
Product Details & Documents
The SlideFolder is manufactured from heat-resistant, injection moulded plastic and can hold up to 20 standard microscope slides.
  • Numbered spaces and transparent leaves for easier identification
  • Five colours allowing colour coded classification of the slides
  • Slides are removed simply by pressing on one end
  • Can be stacked to save space
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    Availability
    Price
  • 720-0479
    Slide holders
    Slide holders
    Assorted
    Plastic
    20
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 271,00 €
    271,00 €
    1 * 10 ST
       271,00 €
  • 720-0480
    Slide holders
    Slide holders
    Blue
    Plastic
    20
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 271,00 €
    271,00 €
    1 * 10 ST
       271,00 €
  • 720-0481
    Slide holders
    Slide holders
    Green
    Plastic
    20
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 271,00 €
    271,00 €
    1 * 10 ST
       271,00 €
  • 720-0482
    Slide holders
    Slide holders
    Grey
    Plastic
    20
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 271,00 €
    271,00 €
    1 * 10 ST
       271,00 €
  • 720-0483
    Slide holders
    Slide holders
    White
    Plastic
    20
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 271,00 €
    271,00 €
    1 * 10 ST
       271,00 €
  • 720-0484
    Slide holders
    Slide holders
    Yellow
    Plastic
    20
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 271,00 €
    271,00 €
    1 * 10 ST
       271,00 €
  • 630-1028
    Slide holders
    Slide holders
    Opaque/white
    Plastic
    20
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 294,00 €
    294,00 €
    1 * 10 ST
       294,00 €