Orders LinkContactUsLinkComponent
VWR®, Cover Slips, Rectangular
VWR®, Cover Slips, Rectangular
VWR®, Cover Slips, Rectangular
Supplier:  VWR Collection
Certificates
VWR®, Cover Slips, Rectangular
VWR®, Cover Slips, Rectangular
5 Options Available Below
Product Details & Documents
These cover slips are specially designed for diagnostic applications.
  • Made of pure white Hydrolytic Class 1 glass
Thickness: 0.13 - 0.16 mm
Thickness: 0,13 - 0,16 mm
Packaging: In dispenser boxes with separate lid.
Product Family Options
Product Information
  • L×W
    No.
    Availability
    Price
  • 21×26 mm
    #1
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 106,00 €
    106,00 €
    1 * 2.000 ST
       106,00 €
  • 24×32 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 61,80 €
    61,80 €
    1 * 1.000 ST
       61,80 €
  • 24×40 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,90 €
    73,90 €
    1 * 1.000 ST
       73,90 €
  • 24×50 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 87,30 €
    87,30 €
    1 * 1.000 ST
       87,30 €
  • 24×60 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 101,00 €
    101,00 €
    1 * 1.000 ST
       101,00 €