Orders LinkContactUsLinkComponent
Rectangular cover glasses, Menzel Gläser
Supplier:   Epredia
Rectangular cover glasses, Menzel Gläser
Supplier:  Epredia
Certificates
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
25 Options Available Below
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles or flaws
  • Made of pre-washed pure white Hydrolytic Class 1 glass
Product Family Options
Product Information
  • L×W
    No.
    Availability
    Price
  • 631-1335
    Cover glasses
    Cover glasses
    21×26 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 48,00 €
    48,00 €
    1 * 1.000 ST
       48,00 €
  • 631-0852
    Cover glasses
    Cover glasses
    22×32 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 57,90 €
    57,90 €
    1 * 1.000 ST
       57,90 €
  • 630-2879
    Cover glasses
    Cover glasses
    22×32 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 66,50 €
    66,50 €
    1 * 1.000 ST
       66,50 €
  • 631-1370
    Cover glasses
    Cover glasses
    22×40 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 82,40 €
    82,40 €
    1 * 1.000 ST
       82,40 €
  • 631-1365
    Cover glasses
    Cover glasses
    22×50 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 83,50 €
    83,50 €
    1 * 1.000 ST
       83,50 €
  • 630-2103
    Cover glasses
    Cover glasses
    22×60 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 94,00 €
    94,00 €
    1 * 1.000 ST
       94,00 €
  • 630-2877
    Cover glasses
    Cover glasses
    22×60 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 109,00 €
    109,00 €
    1 * 1.000 ST
       109,00 €
  • Cover glasses
    24×32 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 59,20 €
    59,20 €
    1 * 1.000 ST
       59,20 €
  • 630-2873
    Cover glasses
    Cover glasses
    24×32 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 68,10 €
    68,10 €
    1 * 1.000 ST
       68,10 €
  • 631-0712
    Cover glasses
    Cover glasses
    24×36 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 74,10 €
    74,10 €
    1 * 1.000 ST
       74,10 €
  • 630-2871
    Cover glasses
    Cover glasses
    24×36 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 85,30 €
    85,30 €
    1 * 1.000 ST
       85,30 €
  • 631-1333
    Cover glasses
    Cover glasses
    24×40 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 72,10 €
    72,10 €
    1 * 1.000 ST
       72,10 €
  • 630-2868
    Cover glasses
    Cover glasses
    24×50 mm
    #0
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 97,50 €
    97,50 €
    1 * 1.000 ST
       97,50 €
  • 630-2867
    Cover glasses
    Cover glasses
    24×50 mm
    #2
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 106,00 €
    106,00 €
    1 * 1.000 ST
       106,00 €
  • 631-9430
    Cover glasses
    Cover glasses
    24×50 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 97,00 €
    97,00 €
    1 * 1.000 ST
       97,00 €
  • 631-9444
    Cover glasses
    Cover glasses
    24×55 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 104,00 €
    104,00 €
    1 * 1.000 ST
       104,00 €
  • 630-2864
    Cover glasses
    Cover glasses
    24×60 mm
    #0
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 113,00 €
    113,00 €
    1 * 1.000 ST
       113,00 €
  • Cover glasses
    24×60 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 97,70 €
    97,70 €
    1 * 1.000 ST
       97,70 €
  • 630-2895
    Cover glasses
    Cover glasses
    24×60 mm
    #2
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 123,00 €
    123,00 €
    1 * 1.000 ST
       123,00 €
  • 631-0853
    Cover glasses
    Cover glasses
    24×60 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 113,00 €
    113,00 €
    1 * 1.000 ST
       113,00 €
  • Cover glasses
    26×76 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 362,00 €
    362,00 €
    1 * 1.000 ST
       362,00 €
  • MENZBC026076AC40
    Cover glasses
    Cover glasses
    26×76 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 415,00 €
    415,00 €
    1 * 1.000 ST
       415,00 €
  • MENZBC040050A1
    Cover glasses
    Cover glasses
    40×50 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 359,00 €
    359,00 €
    1 * 1.000 ST
       359,00 €
  • MENZBC040060A1
    Cover glasses
    Cover glasses
    40×60 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 439,00 €
    439,00 €
    1 * 1.000 ST
       439,00 €
  • 631-9431
    Cover glasses
    Cover glasses
    50×75 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 685,00 €
    685,00 €
    1 * 1.000 ST
       685,00 €