Orders LinkContactUsLinkComponent
Flasks on slides, Nunc™
Flasks on slides, Nunc™
   Sustainable Options Available
Flasks on slides, Nunc™
   Sustainable Options Available
Certificates
Flasks on slides, Nunc™
2 Options Available Below
Product Details & Documents
Glass (Flaskette), PS (SlideFlask), sterile for cell culture on a microscope slide.
  • PS SlideFlask Nunclon™Δ certified
  • SlideFlask is ultrasonically welded to the slide, and individually leak tested
For karyotyping of cells, single cell autoradiography and single cell immuno-fluorescence.
W×D (Flaskette): 20×52 mm; W×D (SlideFlask): 18×50 mm
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2107
  • 732-2609
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
  • Recommended working volume
    Growth area
    Material
    Availability
    Price
  • 734-2107
    SlideFlask (18×50 mm)
    SlideFlask (18×50 mm)
    2,5 − 5 ml
    9 cm²
    PS
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 817,00 €
    817,00 €
    1 * 50 ST
       817,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    734-2107
    Environmentally Preferable
    Environmental certification
  • 732-2609
    Flaskette (20×52 mm)
    Flaskette (20×52 mm)
    2,5 − 5 ml
    10 cm²
    Glass
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 334,00 €
    334,00 €
    1 * 16 ST
       334,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-2609
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact