Order Entry
Austria
ContactUsLinkComponent
 
VWR® Polysine®, Adhesion Slides
  631-0107
undefined
VWR® Polysine®, Adhesion Slides
  631-0107

 

  • Colour:
    White
  • Pk:
    72

 

 

These slides are coated with a permanent adhesive that provides excellent cell and tissue adhesion.

  • Tissue section thickness of 2 to 5 µm
  • 90° ground edges and colour-frosted end for labelling
  • Adhesion superior to glue-, protein- or silane-treated slides

Recommended for molecular biology techniques and cytocentrifuged preparations to reduce tissue loss during prolonged DNA hybridisation and immunocytochemistry.