VWR® SuperFrost® Plus, Adhesion Slides
Supplier: VWR Collection
Certificates
About this item
Superfrost Plus slides are specially treated for frozen tissue sections.
- Permanent positive charge binds freshly frozen tissue sections and cytology preparations
- Ground edges and colour-frosted ends for labelling
- Tissue section thickness: 2 to 5 µm
Electrostatically binds tissue sections to the slide virtually preventing any loss during staining.
Cytology preparations placed on these slides resist cell loss during enzyme digestion, DNA denaturation, and RNA hybridisation.
Frequently Bought Together