Orders LinkContactUsLinkComponent
Adhesion slides, Menzel Gläser, Polysine®
Adhesion slides, Menzel Gläser, Polysine®
Adhesion slides, White
Catalog # 631-1349
Supplier:   Epredia
Adhesion slides, Menzel Gläser, Polysine®
Catalog # 631-1349
Supplier:  Epredia
Adhesion slides, Menzel Gläser, Polysine®
Adhesion slides, Menzel Gläser, Polysine®
Adhesion slides, White
Price
34,20 €
Log in for availability
1 * 72 ST
Product Details & Documents
These slides, made using a technology that electrostatically and chemically attracts fresh cell preparations, formalin-fixed Bouin's or alcohol-fixed paraffin-embedded tissues.
  • 90° ground edges with frosted ends for labelling
  • Adhesion superior to glue-, protein- or silane-treated slides
  • Thick tissue sections of 2 to 5 µm
Unaffected by enzyme pre-digestion or heating, tissue loss is minimised in prolonged in-situ DNA hybridisation techniques and in immunocytochemical methods.
Specifications
  • Colour:
    White
  • Description:
    Adhesion slides