Orders LinkContactUsLinkComponent
PCR racks, 96-well
PCR racks, 96-well
Supplier:   HEATHROW
PCR racks, 96-well
Supplier:  HEATHROW
Certificates
PCR racks, 96-well
PCR racks, 96-well
4 Options Available Below
Product Details & Documents
High resistance PP.
  • Tube wells are easily identified by imprinted numbers and letters
  • Removable, hinged lid
Designed to accommodate 96 individual 0,2 ml tubes or 12 or 8 strips of 8 or 12 tubes.
Colour: Natural, blue, green, yellow or assorted colours (one each of blue, green, pink, yellow and natural).
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • 211-3501
    PCR rack, 96-well
    PCR rack, 96-well
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,80 €
    8,80 €
    1 * 1 ST
       8,80 €
  • 211-3502
    PCR rack, 96-well
    PCR rack, 96-well
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,80 €
    8,80 €
    1 * 1 ST
       8,80 €
  • 211-3504
    PCR rack, 96-well
    PCR rack, 96-well
    Natural
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,80 €
    8,80 €
    1 * 1 ST
       8,80 €
  • 211-3505
    PCR rack, 96-well
    PCR rack, 96-well
    Assorted
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 43,70 €
    43,70 €
    1 * 5 ST
       43,70 €