PCR plates, 96-well, SuperPlate™
PP. Thermo Scientific SuperPlate™ provides four times more rigidity for superior robotic handling. Three versions are available, all with ultra-rigid SBS footprint for compatibility with automated systems, and thin walled round bottom wells for optimal heat transfer and maximum sample recovery.
- Enhanced thermal stability to minimise warping during thermal cycling and heat sealing
- Black lettering for quick referencing of wells
- Maximum well volume 0,2 ml when used with adhesive heat seals
- Skirted version is stackable and compatible with automated systems
- Available in white for qPCR applications
Frequently Bought Together