Plates, Nunc™ OmniTrays
About this item
PS, sterile, with lid. Useful for screening libraries and for DNA hybridisation methods, these 90 ml trays are also suitable for standard Petri dish techniques and bacterial growth.
- Compatible with robotic systems
- Sturdy wall design facilitates handling and stacking
- Can be frozen at −20 °C
External dimensions: 128×86 mm
Recommended working volume: 35 ml
Recommended working volume: 35 ml
2 Options Available Below
Product Details & Documents
PS, sterile, with lid. Useful for screening libraries and for DNA hybridisation methods, these 90 ml trays are also suitable for standard Petri dish techniques and bacterial growth.
- Compatible with robotic systems
- Sturdy wall design facilitates handling and stacking
- Can be frozen at −20 °C
External dimensions: 128×86 mm
Recommended working volume: 35 ml
Recommended working volume: 35 ml
Specifications for Product Family
Specifications
- Catalog No:
- 734-0490
- 732-2735
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
- DescriptionSterileVerfügbarkeitPreis
- Specifications:
Low Manufacturing Impact Mehr
Specifications
Cat. No.734-0490Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Specifications:
Low Manufacturing Impact Mehr
Specifications
Cat. No.732-2735Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact