Storage plates, 96-well, MASTERBLOCK®
PP. The 0,5 ml, 1 ml and 2 ml MASTERBLOCK® microplates are ideal for storing samples or for cultivating bacteria or yeasts.
- Biologically inert, resistant to commonly used solvents (such as DMSO, acetonitrile or acetone) and temperature resistant (−196 °C to +121 °C)
- Uniform external dimensions and tolerances with SBS compatible footprint
- Elevated well walls allows them to be easily sealed with adhesive films, heat seals or CapMats
- Alphanumeric well coding
- Available sterile or non-sterile, and in a choice of colours
- Catalog No:
- 736-0134
- 736-0132
- 736-0131
- 736-0135
- 736-0130
- 736-0155
- GREI780280_50
- 736-0156
- 736-0133
- Model:
- 2 ml MASTERBLOCK®
- 1 ml MASTERBLOCK®
- 1 ml MASTERBLOCK®
- 2 ml MASTERBLOCK®
- 1 ml MASTERBLOCK®
- 0,5 ml MASTERBLOCK®
- 2 ml MASTERBLOCK®
- 0,5 ml MASTERBLOCK®
- 2 ml MASTERBLOCK®
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Well profile:
- V-bottom
- U-bottom
- U-bottom
- V-bottom
- U-bottom
- V-bottom
- V-bottom
- V-bottom
- V-bottom
- Total volume:
- 2,42 ml
- 1,22 ml
- 1,22 ml
- 2,42 ml
- 1,22 ml
- 0,78 ml
- 2,42 ml
- 0,78 ml
- 2,42 ml
- Working volume at -20 °C:
- 0,1 - 2,0
- 0,05 - 1,0
- 0,05 - 1,0
- 0,1 - 2,0
- 0,05 - 1,0
- 0,03 - 0,55
- 0,1 - 2,0
- 0,03 - 0,55
- 0,1 - 2,0
- Working volume at RT:
- 0,1 - 2,3 ml
- 0,05 - 1,2 ml
- 0,05 - 1,2 ml
- 0,1 - 2,3 ml
- 0,05 - 1,2 ml
- 0,03 - 0,7 ml
- 0,1 - 2,1 ml
- 0,03 - 0,7 ml
- 0,1 - 2,3 ml
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Frequently Bought Together