ContactUsLinkComponent
Lids for microplates, Whatman™
Lids for microplates, Whatman™
Catalog # 512-1093
undefined
Lids for microplates, Whatman™
Catalog # 512-1093
Specifications
  • Colour:
    Clear
  • Description:
    Lid, PS, universal
  • Pk:
    100
Specifications
About this item
Universal lids suitable for robotic handling. Can be used as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.