Plates, collagen I coated, Nunc™
PS ready to use plates pre-coated with collagen I, with lid.
- Consistent growth surface from each lot ensures guaranteed performance
- High quality surface promotes cell attachment, growth and differentiation in serum-free and serum-containing media
- Stable at room temperature
W×L: 128×86 mm.
- Catalog No:
- 735-0226
- 734-1187
- 734-1183
- 734-1185
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together