Optical bottom plates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
About this item
PS upper structure with either PS film or no. 1.5 coverglass base. Designed for fluorescence and luminescence based assays, these plates have white or black upper structures with optically clear bottoms. Consequently, they can be read from bottom and top, or both.
- Compatible with automated handling equipment
- Alphanumeric well identification
- Optimised for scintillation counting
- Low fluorescence background ensures accurate results, due to higher signal-to-noise ratios
Specifications
- Catalog No:
- 732-2742
- 734-2180
- 734-2086
- 734-2178
- 732-2741
- 734-2181
- 732-2740
- 734-2032
- 734-2031
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement