Measuring cylinders, tall form, class A, borosilicate glass
Supplier: HIRSCHMANN
Certificates
About this item
DURAN®, borosilicate glass, tall form, class A.
- Calibrated to contain 'ln', certificate of conformity (DE-M marking) with dated batch identification available via website: (www.hirschmannlab.de)
- Hexagonal glass base, with spout
- Amber graduations with ring marks at major graduations
DIN EN ISO 4788
Certifications: DIN EN ISO 4788
Related Information
Certificates
Search by Lot NumberHirschmann brochure EM-Techcolor -volumetric glassware-
Product Literature
Frequently Bought Together