ContactUsLinkComponent
Full face masks, BRK 820
Full face masks, BRK 820
Catalog # 111-3060
Supplier:  BARTELS RIEGER
undefined
Full face masks, BRK 820
Catalog # 111-3060
Supplier:  BARTELS RIEGER
Specifications
  • Type:
    BRK 820, rubber with PC lens
  • Size:
    U*
  • Pk:
    1
Specifications
About this item
This reusable full face mask is for use in industry, agriculture, rescue work and the disposal of hazardous substances. In addition to its use as a sterile filter mask, the mask also functions as a respiration mask for use with lightweight tubing equipment thanks to its standard round thread connection, in accordance with EN 148-1.
  • Visibility is maximised due to the low, space-optimising connectors; viewing pane made from heat-resistant polycarbonate
  • Integrated dampers prevent injury caused by impact
  • With polyester speech diaphragm and five-point head-strap
  • Two exhalation valves ensure low resistance when exhaling; optimal airflow ensures an anti-fog view
  • Choice of skin-friendly, highly durable rubber or silicone, specifically for allergy sufferers
Certifications: EN 136, EN 148
Caution: *One size fits all