Reusable half masks, 7700 series
Supplier: Honeywell Safety
Certificates
About this item
Soft, hypo-allergenic silicone half masks. Interchangeable cartridges and filters provide air-purifying protection for a variety of applications. Direct cartridge-to-face-piece connection eliminates gaskets and cartridge receptacles.
- Soft silicone makes these respirators close fitting and comfortable
- Fit snugly while the cradle suspension system alleviates pressure points
- Can be worn with safety glasses and goggles
Certifications: NIOSH approved for impact and penetration resistance when used with North Safety cartridges and/or filters.
Ordering information: Cartridges and filters must be ordered separately
Frequently Bought Together